Ethyl-4-(3-bromophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

044811-500MG 044811-1G 044811-5G
101867-698EA 278.46 USD
101867-698 101867-700 101867-702
Ethyl-4-(3-bromophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-4-(3-bromophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₆BrNO₃
MW: 338.21 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD00664290
CAS Number: 338960-00-2

Order Now

Specification Test Results

Melting point 172-174°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR