2-(4-Fluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dioxo-2-(4-fluorophenyl)-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

044809-500MG 044809-1G 044809-5G
101867-686EA 556.92 USD
101867-686 101867-688 101867-690
2-(4-Fluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
2-(4-Fluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

Hazard: Irritant

Formula: C₁₀H₅FN₄O₂
MW: 232.18 g/mol
Melting Pt: 242…244 °C
MDL Number: MFCD00794272
CAS Number: 338982-40-4

Order Now

Specification Test Results

Melting point 242-244°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR