2-(3-Methoxyphenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

044806-500MG 044806-1G 044806-5G
101867-668EA 556.92 USD
101867-668 101867-670 101867-672
2-(3-Methoxyphenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile ≥95%
2-(3-Methoxyphenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

Hazard: Irritant

Formula: C₁₁H₈N₄O₃
MW: 244.21 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD00794268
CAS Number: 338982-38-0

Order Now

Specification Test Results

Melting point 207-209°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR