2-Chloro-N-[4-(2-chlorophenyl)-1,3-thiazol-2-yl]acetamide ≥95%

Supplier: Matrix Scientific

044802-500MG 044802-1G 044802-5G
101867-662EA 278.46 USD
101867-662 101867-664 101867-666
2-Chloro-N-[4-(2-chlorophenyl)-1,3-thiazol-2-yl]acetamide ≥95%
2-Chloro-N-[4-(2-chlorophenyl)-1,3-thiazol-2-yl]acetamide

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂OS
Melting Pt: 130…132 °C
MDL Number: MFCD01315353
CAS Number: 338957-50-9

Order Now

Specification Test Results

Melting point 130-132°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR