2-[2-(2-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid ≥95%

Supplier: Matrix Scientific

044799-500MG 044799-1G 044799-5G
101867-656EA 747.69 USD
101867-656 101867-658 101867-660
2-[2-(2-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid ≥95%
2-[2-(2-Chlorophenyl)-4-phenyl-1,3-thiazol-5-yl]acetic acid

Hazard: Irritant

Formula: C₁₇H₁₂ClNO₂S
Melting Pt: 174…176 °C
MDL Number: MFCD00664027
CAS Number: 23821-79-6

Order Now

Specification Test Results

Melting point 174-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR