4-(Chloromethyl)-5-[(2-chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044798-500MG 044798-1G 044798-5G
101867-650EA 921.54 USD
101867-650 101867-652 101867-654
4-(Chloromethyl)-5-[(2-chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole ≥95%
4-(Chloromethyl)-5-[(2-chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₇H₁₄Cl₂N₂S
Melting Pt: 69…70 °C
MDL Number: MFCD00664820
CAS Number: 318289-54-2

Order Now

Specification Test Results

Melting point 69-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR