2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile ≥95%

Supplier: Matrix Scientific

044796-500MG 044796-1G 044796-5G
101867-638EA 400 USD
101867-638 101867-640 101867-642
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile ≥95%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]malononitrile

Hazard: Irritant

Formula: C₉H₃ClF₃N₃
MW: 245.59 g/mol
Melting Pt: 210 °C
MDL Number: MFCD01315462
CAS Number: 338956-33-5

Order Now

Specification Test Results

Melting point 210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR