2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: (2Z)-2-(3-Chlorophenyl)-3-(dimethylamino)prop-2-enal

044793-500MG 044793-1G 044793-5G
101867-626EA 278.46 USD
101867-626 101867-628 101867-630
2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde ≥95%
2-(3-Chlorophenyl)-3-(dimethylamino)acrylaldehyde

Hazard: Irritant

Formula: C₁₁H₁₂ClNO
MW: 209.68 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD04125587
CAS Number: 86723-63-9

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR