2-Chloro-5-(4-chlorophenyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

044792-500MG 044792-1G 044792-5G
101867-620EA 747.69 USD
101867-620 101867-622 101867-624
2-Chloro-5-(4-chlorophenyl)nicotinonitrile ≥95%
2-Chloro-5-(4-chlorophenyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₆Cl₂N₂
Melting Pt: 182…184 °C
MDL Number: MFCD00231583
CAS Number: 35982-99-1

Order Now

Specification Test Results

Melting point 182-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR