Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

044791-1G 044791-5G
101867-616EA 769.23 USD
101867-616 101867-618
Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀BrNO₃
MW: 296.1200 g/mol
Melting Pt: 326-329°C
MDL Number: MFCD00173362
CAS Number: 122794-99-4

Order Now

Specification Test Results

Melting point 326-329°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR