Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

044790-500MG 044790-1G 044790-5G
101867-608EA 278.46 USD
101867-608 101867-610 101867-612
Ethyl-6-fluoro-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀FNO₃
MW: 235.22 g/mol
Melting Pt: 320-323°C
MDL Number: MFCD00173345
CAS Number: 318-35-4

Order Now

Specification Test Results

Melting point 320-323°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR