2-(5-Acetyl-2-thienyl)acetonitrile ≥95%

Supplier: Matrix Scientific

044789-500MG 044789-1G 044789-5G
101867-602EA 278.46 USD
101867-602 101867-604 101867-606
2-(5-Acetyl-2-thienyl)acetonitrile ≥95%
2-(5-Acetyl-2-thienyl)acetonitrile

Hazard: Irritant

Formula: C₈H₇NOS
Melting Pt: 47…49 °C
MDL Number: MFCD00665058
CAS Number: 107701-61-1

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR