2-{[2-(4-Methoxyanilino)-2-oxoethyl]sulfonyl}acetic acid ≥95%

Supplier: Matrix Scientific

044784-500MG 044784-1G 044784-5G
101867-586EA 181.54 USD
101867-586 101867-588 101867-590
2-{[2-(4-Methoxyanilino)-2-oxoethyl]sulfonyl}acetic acid ≥95%
2-{[2-(4-Methoxyanilino)-2-oxoethyl]sulfonyl}acetic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₆S
Melting Pt: 201…203 °C
MDL Number: MFCD00202414
CAS Number: 338953-92-7

Order Now

Specification Test Results

Melting point 201-203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR