4-Phenyl-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

044783-500MG 044783-1G 044783-5G
101867-580EA 100 USD
101867-580 101867-582 101867-584
4-Phenyl-1H-pyrazol-5-amine ≥95%
4-Phenyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₉H₉N₃
Melting Pt: 175…177 °C
MDL Number: MFCD00231797
CAS Number: 57999-06-1

Order Now

Specification Test Results

Melting point 175-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR