2-Chloro-6-[(4-chlorophenyl)(cyano)methyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

044782-500MG 044782-1G 044782-5G
101867-574EA 181.54 USD
101867-574 101867-576 101867-578
2-Chloro-6-[(4-chlorophenyl)(cyano)methyl]benzenecarbonitrile ≥95%
2-Chloro-6-[(4-chlorophenyl)(cyano)methyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₅H₈Cl₂N₂
Melting Pt: 104…106 °C
MDL Number: MFCD00231962
CAS Number: 338953-31-4

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR