Ethyl-4-(2-pyridinylcarbonyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044781-500MG 044781-1G
101867-570EA 747.69 USD
101867-570 101867-572
Ethyl-4-(2-pyridinylcarbonyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(2-pyridinylcarbonyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄
Melting Pt: 61…63 °C
MDL Number: MFCD00664774
CAS Number: 338761-70-9

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR