Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044779-500MG 044779-1G
101867-560EA 1109.23 USD
101867-560 101867-562
Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate ≥95%
Ethyl-5-methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₃F₃N₂O₄
MW: 342.28 g/mol
Melting Pt: 103…105 °C
MDL Number: MFCD00664767
CAS Number: 338761-34-5

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR