Ethyl-4-[(4-chloroanilino)carbonyl]-5-methyl-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044777-500MG 044777-1G
101867-550EA 1109.23 USD
101867-550 101867-552
Ethyl-4-[(4-chloroanilino)carbonyl]-5-methyl-3-isoxazolecarboxylate ≥95%
Ethyl-4-[(4-chloroanilino)carbonyl]-5-methyl-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O₄
Melting Pt: 108…110 °C
MDL Number: MFCD00664765
CAS Number: 134889-04-6

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR