α-Bromo-2-chloro-5-nitrotoluene ≥95%

Supplier: Matrix Scientific

044776-500MG 044776-1G 044776-5G
101867-544EA 269.23 USD
101867-544 101867-546 101867-548
α-Bromo-2-chloro-5-nitrotoluene ≥95%
α-Bromo-2-chloro-5-nitrotoluene

Hazard: Irritant

Formula: C₇H₅BrClNO₂
Melting Pt: 85-87°C
CAS Number: 52427-01-7

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR