5-[(4-Chlorophenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

044775-500MG 044775-1G
101867-540EA 700 USD
101867-540 101867-542
5-[(4-Chlorophenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylic acid ≥95%
5-[(4-Chlorophenyl)sulfanyl]-1,2,3-thiadiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₅ClN₂O₂S₂
Melting Pt: 189…190 °C
MDL Number: MFCD00794805
CAS Number: 338760-78-4

Order Now

Specification Test Results

Melting point 189-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR