[5-[(4-Chlorobenzyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%

Supplier: Matrix Scientific

044772-500MG 044772-1G 044772-5G
101867-526EA 921.54 USD
101867-526 101867-528 101867-530
[5-[(4-Chlorobenzyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[5-[(4-Chlorobenzyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₃H₁₂ClF₃N₂OS
MW: 336.77 g/mol
Melting Pt: 67…70 °C
MDL Number: MFCD00172667
CAS Number: 318469-47-5

Order Now

Specification Test Results

Melting point 67-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR