{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol ≥95%

Supplier: Matrix Scientific

044771-500MG 044771-1G 044771-5G
101867-520EA 747.69 USD
101867-520 101867-522 101867-524
{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol ≥95%
{1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl}methanol

Hazard: Irritant

Formula: C₁₃H₁₀F₆N₂O₂
MW: 340.23 g/mol
Melting Pt: 81…83 °C
MDL Number: MFCD00172663
CAS Number: 318469-40-8

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR