[5-[(4-Fluorophenyl)sulfonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(4-Fluorobenzenesulphonyl)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole

044769-500MG 044769-1G 044769-5G
101867-508EA 921.54 USD
101867-508 101867-510 101867-512
[5-[(4-Fluorophenyl)sulfonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[5-[(4-Fluorophenyl)sulfonyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₂H₁₀F₄N₂O₃S
MW: 338.28 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00172654
CAS Number: 318469-24-8

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR