[1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(Hydroxymethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
044768-500MG
044768-1G
044768-5G
101867-502EA
747.69
USD
101867-502
101867-504
101867-506
[1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₂H₁₁F₃N₂O₂ MW: 272.23 g/mol Melting Pt: 80…82 °C |
MDL Number:
MFCD00172652 CAS Number: 318469-22-6 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...