1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044767-500MG 044767-1G 044767-5G
101867-496EA 747.69 USD
101867-496 101867-498 101867-500
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₂
MW: 270.21 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD01936001
CAS Number: 109925-42-0

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR