Ethyl-3-amino-3-(4-methoxyanilino)acrylate hydrochloride ≥95%

Supplier: Matrix Scientific

044763-500MG 044763-1G 044763-5G
101867-478EA 375.38 USD
101867-478 101867-480 101867-482
Ethyl-3-amino-3-(4-methoxyanilino)acrylate hydrochloride ≥95%
Ethyl-3-amino-3-(4-methoxyanilino)acrylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₃
Melting Pt: 155…157 °C
MDL Number: MFCD00243965
CAS Number: 318469-17-9

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR