5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

044762-500MG 044762-1G 044762-5G
101867-472EA 556.92 USD
101867-472 101867-474 101867-476
5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid ≥95%
5-Bromo-2-oxo-1-[3-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉BrF₃NO₃
MW: 376.14 g/mol
Melting Pt: 187…188 °C
MDL Number: MFCD01935940
CAS Number: 338982-36-8

Order Now

Specification Test Results

Melting point 187-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR