1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044760-500MG 044760-1G 044760-5G
101867-466EA 1023.08 USD
101867-466 101867-468 101867-470
1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde ≥95%
1-Phenyl-1H-1,2,3-triazole-4-carbaldehyde

Hazard: Irritant

Formula: C₉H₇N₃O
Melting Pt: 99…101 °C
MDL Number: MFCD00100219
CAS Number: 34296-51-0

Order Now

Specification Test Results

Melting point 99-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR