1-(2-Ethoxy-2-oxoethyl)pyridinium bromide ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2-Ethoxy-2-oxoethyl)pyridin-1-ium bromide

044758-1G 044758-5G
101867-462EA 150.77 USD
101867-462 101867-464
1-(2-Ethoxy-2-oxoethyl)pyridinium bromide ≥95%
1-(2-Ethoxy-2-oxoethyl)pyridinium bromide

Hazard: Irritant

Formula: C₉H₁₂BrNO₂
Melting Pt: 137…139 °C
MDL Number: MFCD00463694
CAS Number: 17282-40-5

Order Now

Specification Test Results

Melting point 137-139°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR