1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(3,4-Dichlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

044753-500MG 044753-1G 044753-5G
101867-440EA 278.46 USD
101867-440 101867-442 101867-444
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
1-(3,4-Dichlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₃
MW: 298.13 g/mol
Melting Pt: 253…256.5 °C
MDL Number: MFCD00139865
CAS Number: 338755-21-8

Order Now

Specification Test Results

Melting point 253-256.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR