1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3(2H)-one

044751-500MG 044751-1G 044751-5G
101867-430EA 181.54 USD
101867-430 101867-432 101867-434
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol ≥95%
1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-ol

Hazard: Irritant

Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
Melting Pt: 150…151.5 °C
MDL Number: MFCD00085094
CAS Number: 119022-51-4

Order Now

Specification Test Results

Melting point 150-151.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR