1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

044750-500MG 044750-1G 044750-5G
101867-424EA 209.23 USD
101867-424 101867-426 101867-428
1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-Benzyl-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₃
Melting Pt: 133.5…134.5 °C
MDL Number: MFCD00139999
CAS Number: 89960-36-1

Order Now

Specification Test Results

Melting point 133.5-134.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR