1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(3-Chlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

044749-500MG 044749-1G 044749-5G
101867-418EA 278.46 USD
101867-418 101867-420 101867-422
1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-(3-Chlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₃
MW: 263.68 g/mol
Melting Pt: 187…188.5 °C
MDL Number: MFCD00139984
CAS Number: 338754-68-0

Order Now

Specification Test Results

Melting point 187-188.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR