1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole

044746-500MG 044746-1G 044746-5G
101867-406EA 278.46 USD
101867-406 101867-408 101867-410
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol ≥95%
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol

Hazard: Irritant

Formula: C₅H₅F₃N₂O
MW: 166.1 g/mol
Melting Pt: 178…180 °C
MDL Number: MFCD00139833
CAS Number: 122431-37-2

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR