1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(2,6-Dichlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

044745-500MG 044745-1G 044745-5G
101867-400EA 93.85 USD
101867-400 101867-402 101867-404
1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₃
MW: 298.13 g/mol
Melting Pt: 190…192 °C
MDL Number: MFCD00139966
CAS Number: 338754-23-7

Order Now

Specification Test Results

Melting point 190-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR