Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1,2-dihydro-2-oxopyridine-3-carboxylate, 1,2-Dihydro-3-(methoxycarbonyl)-2-oxopyridine

044742-500MG 044742-1G 044742-5G
101867-382EA 30.77 USD
101867-382 101867-384 101867-386
Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate ≥95%
Methyl 2-oxo-1,2-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.137 g/mol
Melting Pt: 115…118 °C
MDL Number: MFCD00661282
CAS Number: 10128-91-3

Order Now

Specification Test Results

Melting point 115-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR