1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-(4-Chlorobenzyl)hydroxylamine hydrochloride
044741-500MG
044741-1G
044741-5G
101867-376EA
78.46
USD
101867-376
101867-378
101867-380
1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-4-chlorobenzene chloride
Hazard: Irritant
Formula:
C₇H₉Cl₂NO MW: 194.0585 g/mol Melting Pt: 230°C |
MDL Number:
MFCD01114589 CAS Number: 38936-60-6 |
Specification Test Results
Melting point | 230°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...