4-[(Ammoniooxy)methyl]-1,2-dichlorobenzene chloride ≥95%
Supplier: Matrix Scientific
044740-500MG
044740-1G
044740-5G
101867-370EA
375.38
USD
101867-370
101867-372
101867-374
4-[(Ammoniooxy)methyl]-1,2-dichlorobenzene chloride ≥95%
4-[(Ammoniooxy)methyl]-1,2-dichlorobenzene chloride
Hazard: Irritant
Formula:
C₇H₈Cl₃NO Melting Pt: 195°C |
MDL Number:
MFCD01114587 CAS Number: 15256-10-7 |
Specification Test Results
Melting point | 195°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...