N-Acetyl-2-cyano-3-(dimethylamino)acrylamide ≥95%

Supplier: Matrix Scientific

044737-500MG 044737-1G 044737-5G
101867-352EA 375.38 USD
101867-352 101867-354 101867-356
N-Acetyl-2-cyano-3-(dimethylamino)acrylamide ≥95%
N-Acetyl-2-cyano-3-(dimethylamino)acrylamide

Hazard: Irritant

Formula: C₈H₁₁N₃O₂
Melting Pt: 126…128 °C
MDL Number: MFCD01315455
CAS Number: 338753-65-4

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR