Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

044736-500MG 044736-1G 044736-5G
101867-346EA 747.69 USD
101867-346 101867-348 101867-350
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-acetyl-1-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD00141903
CAS Number: 85795-19-3

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR