2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-α-phenyl-3-pyridazineacetonitrile, 2-(6-Chloropyridazin-3-yl)-2-phenylacetonitrile

044734-500MG 044734-1G 044734-5G
101867-334EA 181.54 USD
101867-334 101867-336 101867-338
2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile

Hazard: Irritant

Formula: C₁₂H₈ClN₃
MW: 229.6650 g/mol
Melting Pt: 128-130°C
MDL Number: MFCD00140699
CAS Number: 73535-73-6

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR