2-(6-Chloro-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

044733-500MG 044733-1G 044733-5G
101867-328EA 181.54 USD
101867-328 101867-330 101867-332
2-(6-Chloro-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitrile ≥95%
2-(6-Chloro-3-pyridazinyl)-2-(4-methoxyphenyl)acetonitrile

Hazard: Irritant

Formula: C₁₃H₁₀ClN₃O
Melting Pt: 130-132°C
MDL Number: MFCD00140696
CAS Number: 338752-84-4

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR