1-Methyl-2-(trichloroacetyl)pyrrole ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trichloro-1-(1-methyl-1H-pyrrol-2-yl)ethanone

044732-500MG 044732-1G 044732-5G
101867-322EA 278.46 USD
101867-322 101867-324 101867-326
1-Methyl-2-(trichloroacetyl)pyrrole ≥95%
1-Methyl-2-(trichloroacetyl)pyrrole

Hazard: Irritant

Formula: C₇H₆Cl₃NO
MW: 226.4876 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD00052521
CAS Number: 21898-65-7

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR