Methyl-4-hydroxy-1-(4-methylphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Supplier: Matrix Scientific
044730-500MG
044730-1G
044730-5G
101867-310EA
300
USD
101867-310
101867-312
101867-314
Methyl-4-hydroxy-1-(4-methylphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-4-hydroxy-1-(4-methylphenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₄ Melting Pt: 153…155 °C |
MDL Number:
MFCD00141004 CAS Number: 338751-70-5 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...