4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid ≥95%

Supplier: Matrix Scientific

044729-500MG 044729-1G 044729-5G
101867-304EA 747.69 USD
101867-304 101867-306 101867-308
4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid ≥95%
4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₇F₃N₂O₄
MW: 300.2 g/mol
Melting Pt: 261…263 °C
MDL Number: MFCD00141002
CAS Number: 121582-64-7

Order Now

Specification Test Results

Melting point 261-263°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR