3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide ≥95%

Supplier: Matrix Scientific

044727-500MG 044727-1G 044727-5G
101867-294EA 375.38 USD
101867-294 101867-296 101867-298
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide ≥95%
3-[(4-Fluorobenzyl)oxy]-2-thiophenecarbohydrazide

Hazard: Irritant

Formula: C₁₂H₁₁FN₂O₂S
MW: 266.3 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD00215161
CAS Number: 338751-50-1

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR