2-{1-Amino-2-[(4-chlorophenyl)sulfanyl]ethylidene}malononitrile ≥95%

Supplier: Matrix Scientific

044724-500MG 044724-1G 044724-5G
101867-282EA 375.38 USD
101867-282 101867-284 101867-286
2-{1-Amino-2-[(4-chlorophenyl)sulfanyl]ethylidene}malononitrile ≥95%
2-{1-Amino-2-[(4-chlorophenyl)sulfanyl]ethylidene}malononitrile

Hazard: Irritant

Formula: C₁₁H₈ClN₃S
Melting Pt: 215…217 °C
MDL Number: MFCD00139196
CAS Number: 338750-88-2

Order Now

Specification Test Results

Melting point 215-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR