2-[Amino(3-pyridinyl)methylene]malononitrile ≥95%

Supplier: Matrix Scientific

044722-500MG 044722-1G 044722-5G
101867-270EA 181.54 USD
101867-270 101867-272 101867-274
2-[Amino(3-pyridinyl)methylene]malononitrile ≥95%
2-[Amino(3-pyridinyl)methylene]malononitrile

Hazard: Irritant

Formula: C₉H₆N₄
MW: 170.17 g/mol
Melting Pt: 211…214 °C
MDL Number: MFCD00139195
CAS Number: 124883-64-3

Order Now

Specification Test Results

Melting point 211-214°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR