2-[Amino(4-pyridinyl)methylene]malononitrile ≥95%

Supplier: Matrix Scientific

044721-500MG 044721-1G 044721-5G
101867-264EA 181.54 USD
101867-264 101867-266 101867-268
2-[Amino(4-pyridinyl)methylene]malononitrile ≥95%
2-[Amino(4-pyridinyl)methylene]malononitrile

Hazard: Irritant

Formula: C₉H₆N₄
MW: 170.17 g/mol
Melting Pt: 228…230 °C
MDL Number: MFCD00139194
CAS Number: 338750-87-1

Order Now

Specification Test Results

Melting point 228-230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR