(1-Amino-2-chloroethylidene)malononitrile ≥95%

Supplier: Matrix Scientific

044720-500MG 044720-1G 044720-5G
101867-258EA 181.54 USD
101867-258 101867-260 101867-262
(1-Amino-2-chloroethylidene)malononitrile ≥95%
(1-Amino-2-chloroethylidene)malononitrile

Hazard: Irritant

Formula: C₅H₄ClN₃
Melting Pt: 157 °C
CAS Number: 118645-77-5

Order Now

Specification Test Results

Melting point 157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR